Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Disodium Salt, MP Biomedicals™

Catalog No. ICN19518425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19518425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19518425 Supplier MP Biomedicals, Inc Supplier No. 0219518425

missing translation for 'validMainframeMsg'

Disodium salt of fluorescein, a biodegradable dye and a noted pH sensitive fluorophore. Utilized in studies as a drug model

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-47-8
Molecular Formula C20H10Na2O5
MDL Number MFCD00167039
Synonym Soluble Fluorescein, Aizen Uranine
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Weight (g/mol) 376.28
Formula Weight 376.28
Color Orange
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fluorescein Disodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.