Learn More
CELLMOSAIC INC FLUORESCEIN-DOXORUBICIN CN 5MG
SDP
Supplier:  CELLMOSAIC INC CM621025MG
Encompass
Chemical name: Fluorescein-Doxorubicin conjugateChemical formula: C48H40N2O16SMolecular weight: 932.91 CAS number: N/APhysical appearance: orange powderStorage temp.: -20 °C Purity: ≥ 93% by HPLC
Catalog No. NC2542615