Learn More
Jackson Immuno Research Labs Fluorescein (FITC) AffiniPure® F(ab')₂ Fragment Donkey Anti-Human IgG (H+L) 0.3 mg
SDP
Catalog No. NC3971049
Encompass_Preferred
Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule human IgG. It also reacts with the light chains of other human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, chicken, goat, guinea pig, syrian hamster, horse, mouse, rabbit, rat and sheep serum proteins, but it may cross-react with immunoglobulins from other species.
Catalog No. NC3971049 Supplier Jackson Immuno Research Labs Supplier No. 709096149
Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule human IgG. It also reacts with the light chains of other human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, chicken, goat, guinea pig, syrian hamster, horse, mouse, rabbit, rat and sheep serum proteins, but it may cross-react with immunoglobulins from other species.