Learn More
Abcam Fluorescein di-ß-D-glucopyranoside (FDGlu), 5MG
Catalog No. 50996178
Encompass_Preferred
MW 656.6 Da, Purity >95%. Ultrasensitive fluorogenic β-glucosidase substrate. Releases fluorescent dye fluorescein (Ex /Em = 490/514 nm) in direct correlation with enzyme activity *in vivo* or *in vitro*. Used for quantification of enzyme activity and cell viability determination.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-178 Supplier Abcam Supplier No. AB2738935MG
MW 656.6 Da, Purity >95%. Ultrasensitive fluorogenic β-glucosidase substrate. Releases fluorescent dye fluorescein (Ex /Em = 490/514 nm) in direct correlation with enzyme activity *in vivo* or *in vitro*. Used for quantification of enzyme activity and cell viability determination.
The product is subject to the following: Abcam Restricted Use Statement