Learn More
Biotium Fluorescein hexylamine
Catalog No. 501964835
Encompass_Preferred
Fluorescein hexylamine can be coupled to activated carboxy groups or sulfonyl chlorides.  This product could be used as a direct replacement to fluorescein cadaverine. Ex / Em (pH9) = 493/517 nm Yellow solid soluble in DMSO, DMF, or pH>6.5 buffers Store at 4°C and protect from light C 29 H 27 F 3 N 2 O 8 MW: 588.54
Catalog No. 50-196-4835 Supplier Biotium Supplier No. 91058
Fluorescein hexylamine can be coupled to activated carboxy groups or sulfonyl chlorides.  This product could be used as a direct replacement to fluorescein cadaverine. Ex / Em (pH9) = 493/517 nm Yellow solid soluble in DMSO, DMF, or pH>6.5 buffers Store at 4°C and protect from light C 29 H 27 F 3 N 2 O 8 MW: 588.54