Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Fluorescein Isothiocyanate, Isomer I 0.5 GR
SDP

Catalog No. 50980477
Encompass_Preferred

CAS #3326-32-7 C₂₁H₁₁NO₅S F.W. 389.39
Solubility: Water soluable
Assay: >90%

Fluorescein Isothiocyanate, Isomer I, Certified (FITC) is used in Culing's Method for fluorescent antibody staining for demonstration of specific antigens. Water soluble.

Catalog No. 50-980-477 Supplier Electron Microscopy Sciences Supplier No. 15670
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #3326-32-7 C₂₁H₁₁NO₅S F.W. 389.39
Solubility: Water soluable
Assay: >90%

Fluorescein Isothiocyanate, Isomer I, Certified (FITC) is used in Culing's Method for fluorescent antibody staining for demonstration of specific antigens. Water soluble.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.