Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Isothiocyanate Isomer I, ≥97.5%, Sigma-Aldrich

Catalog No. 01258871 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-871 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-871 Supplier MilliporeSigma Supelco Supplier No. F4274250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bottomless glass bottle. Contents are inside inserted fused cone.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-32-7

Specifications

Format Powder
Quantity 250 mg
Product Type Fluorescence Reagent
Molecular Weight (g/mol) 389.38 g/mol
Purity ≥97.5%
Label Type Other Labels or Dyes
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.