Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Isothiocyanate (mixture of 5- and 6- isomers) 97.0+%, TCI America™
SDP

Catalog No. F0784100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0784100MG 100 mg
F07841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0784100MG Supplier TCI America Supplier No. F0784100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27072-45-3
Molecular Formula C21H11NO5S
MDL Number MFCD00005063
Synonym Isothiocyanatofluorescein

Specifications

Quantity 100 mg
Formula Weight 389.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein Isothiocyanate (mixture of 5- and 6- isomers)
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.