Learn More
Vector Laboratories Lens Culinaris Agglutinin (LCA) Fluorescein-labeled 5 mg
SDP
Supplier:  Vector Laboratories FL10415
Encompass
Lens culinaris agglutinin (LCA) contains two subunits of 17 kDa and two of 8 kDa. It has a narrower specificity than Con A because it recognizes sequences containing α-linked mannose residues and other sugars as part of the receptor structure. An α-linked fucose attached to the N-acetylchitobiose of the core oligosaccharide enhances affinity. The narrower specificity allows subfractionation of glycoproteins and peptides after initial isolation with Con A. LCA prevents skin allograft rejection and can purify numerous glycoproteins, including immunoglobulins, histocompatibility antigens, and α2-macroglobulin. This conjugate features a ratio of fluorophores to lectin protein that provides optimal staining (excitation 495 nm, emission 515 nm). Supplied as a solution essentially free of unconjugated fluorophores, it is preserved with sodium azide. The recommended inhibiting/eluting sugar is a mixture of 200 mM α-methylmannoside/200 mM α-methylglucoside.
Catalog No. NC0182465