Learn More
Medchemexpress LLC Fluorescein-NAD+ | 66.2% | 81 UG
SDP
Catalog No. 5000341125
Encompass_Preferred
Fluorescein-NAD+ is an alternative to radiolabeled NAD and serves as a substrate for ADP-ribosylation. It can be utilized in PARP assays through fluorescence microscopy.
  • Alternative to radiolabeled NAD
  • Substrate for ADP-ribosylation
  • Utilized in PARP assays
  • Used through fluorescence microscopy
  • Extinction coefficient: 262 nm
Catalog No. 50-003-41125 Supplier Medchemexpress LLC Supplier No. HY13100981UG
Fluorescein-NAD+ is an alternative to radiolabeled NAD and serves as a substrate for ADP-ribosylation. It can be utilized in PARP assays through fluorescence microscopy.
  • Alternative to radiolabeled NAD
  • Substrate for ADP-ribosylation
  • Utilized in PARP assays
  • Used through fluorescence microscopy
  • Extinction coefficient: 262 nm