Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fluorescein o-acrylate

Catalog No. 568856500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-885-6500MG 500 mg
56-885-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-885-6500MG Supplier Sigma Aldrich Supplier No. 568856500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fluorescein o-acrylate

Specifications

CAS 193419-86-2
Melting Point 231°C to 233°C
Molecular Formula C23H14O6
Linear Formula C23H14O6
Quantity 500 mg
Synonym 3 -Acryloxyspirobenzo[c]-furan[1,9 ]xanthen-3-one; Acryloyloxy fluorescein
Molecular Weight (g/mol) 386.35
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.