Learn More
Abcam Fluorescein octadecyl ester, Lipophilic fluorescent pH indicator, 500MG
Supplier:  Abcam AB145431500MG
Encompass_Preferred
MW 584.78 g/mol, Purity >97%. Lipophilic fluorescent pH indicator. Also used in an alcohol-selective electrode.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5198