Learn More
Abcam Fluorescein octadecyl ester, Lipophilic fluorescent pH indicator, 50MG
Catalog No. 502705197
Encompass_Preferred
MW 584.78 g/mol, Purity >97%. Lipophilic fluorescent pH indicator. Also used in an alcohol-selective electrode.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5197 Supplier Abcam Supplier No. AB14543150MG
MW 584.78 g/mol, Purity >97%. Lipophilic fluorescent pH indicator. Also used in an alcohol-selective electrode.
The product is subject to the following: Abcam Restricted Use Statement