Learn More
AdipoGen Fluorescein octadecyl ester
SDP
Catalog No. 502560902
Encompass_Preferred
Chemical. CAS 138833-46-2. Formula C38H48O5. MW 584.78. Synthetic. Chromoionophore XI is used as a lipophilic fluorescent pH indicator.
Catalog No. 50-256-0902 Supplier AdipoGen Supplier No. CDXF0074M500
Chemical. CAS 138833-46-2. Formula C38H48O5. MW 584.78. Synthetic. Chromoionophore XI is used as a lipophilic fluorescent pH indicator.