Learn More
Medchemexpress LLC Fluorescein-PEG3-amine | 1807539-04-3 | 96.7% | C29H31N3O8S | 25 MG
SDP
Catalog No. 5000352628
Encompass_Preferred
Fluorescein-PEG3-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, facilitating the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-52628 Supplier Medchemexpress LLC Supplier No. 25MGHY141080
Fluorescein-PEG3-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, facilitating the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins