Learn More
Medchemexpress LLC Fluorescein-PEG3-amine | 1807539-04-3 | MFCD23726633 | 96.7% | C29H31N3O8S | 10MG
SDP
Catalog No. 5000261199
Encompass_Preferred
Fluorescein-PEG3-amine is a PEG-based PROTAC linker comprising a fluorescein fluorophore connected through a three-unit polyethylene glycol spacer to a terminal primary amine. It is supplied as a solid for research use and is intended for conjugation reactions in chemical biology and PROTAC synthesis; follow frozen storage recommendations for long-term stability.
  • Fluorescent probe for visualization and tracking.
  • Terminal primary amine enables straightforward conjugation.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Supplied as a stable powder with long-term frozen storage.
  • Suitable for use as a linker in PROTAC synthesis and bioconjugation workflows.
Catalog No. 50-002-61199 Supplier Medchemexpress LLC Supplier No. HY14108010MG
Fluorescein-PEG3-amine is a PEG-based PROTAC linker comprising a fluorescein fluorophore connected through a three-unit polyethylene glycol spacer to a terminal primary amine. It is supplied as a solid for research use and is intended for conjugation reactions in chemical biology and PROTAC synthesis; follow frozen storage recommendations for long-term stability.
  • Fluorescent probe for visualization and tracking.
  • Terminal primary amine enables straightforward conjugation.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Supplied as a stable powder with long-term frozen storage.
  • Suitable for use as a linker in PROTAC synthesis and bioconjugation workflows.