Learn More
Medchemexpress LLC Fluorescein-PEG4-acid | 1807518-76-8 | 96.3% | 654.68 | 10 MG
SDP
Catalog No. 5000436199
Encompass_Preferred
Fluorescein-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-36199 Supplier Medchemexpress LLC Supplier No. HY12036810MG
Fluorescein-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only