Learn More
Medchemexpress LLC Fluorescein-PEG4-acid | 1807518-76-8 | 96.3% | 654.68 | 25 MG
SDP
Catalog No. 5000439181
Encompass_Preferred
Fluorescein-PEG4-acid is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
Catalog No. 50-004-39181 Supplier Medchemexpress LLC Supplier No. 25MGHY120368
Fluorescein-PEG4-acid is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs