Learn More
Medchemexpress LLC Fluorescein-PEG4-acid | 1807518-76-8 | 96.3% | 654.68 | 250 MG
SDP
Catalog No. 5000439714
Encompass_Preferred
Fluorescein-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two ligands, one for an E3 ubiquitin ligase and another for the target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-004-39714 Supplier Medchemexpress LLC Supplier No. 250MGHY120368
Fluorescein-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two ligands, one for an E3 ubiquitin ligase and another for the target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Enables synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system to selectively degrade target proteins