Learn More
Medchemexpress LLC Fluorescein-PEG4-acid | 1807518-76-8 | 96.3% | 654.68 | 50 MG
SDP
Catalog No. 5000437296
Encompass_Preferred
Fluorescein-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
Catalog No. 50-004-37296 Supplier Medchemexpress LLC Supplier No. HY12036850MG
Fluorescein-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.