Learn More
Medchemexpress LLC Fluorescein-PEG4-acid | 1807518-76-8 | MFCD23726637 | 95.0% | 654.68 g/mol | C32H34N2O11S | 5 MG
SDP
Catalog No. 5000264431
Encompass_Preferred
Fluorescein-PEG4-acid is a PEG-based linker that incorporates a fluorescein dye and terminates in a free carboxylic acid. It is intended for use as a linker in PROTAC synthesis and for covalent conjugation to biomolecules to provide fluorescent labeling for assays and imaging.
  • PEG4 spacer with a fluorescein dye for fluorescence tagging.
  • Terminal carboxylic acid enables amide coupling to amines.
  • Suitable as a PROTAC linker and for fluorescent labeling applications.
  • Yellow to orange solid; molecular weight 654.68 g/mol.
  • Store under nitrogen, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
Catalog No. 50-002-64431 Supplier Medchemexpress LLC Supplier No. HY1203685MG
Fluorescein-PEG4-acid is a PEG-based linker that incorporates a fluorescein dye and terminates in a free carboxylic acid. It is intended for use as a linker in PROTAC synthesis and for covalent conjugation to biomolecules to provide fluorescent labeling for assays and imaging.
  • PEG4 spacer with a fluorescein dye for fluorescence tagging.
  • Terminal carboxylic acid enables amide coupling to amines.
  • Suitable as a PROTAC linker and for fluorescent labeling applications.
  • Yellow to orange solid; molecular weight 654.68 g/mol.
  • Store under nitrogen, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).