Learn More
Biotium Fluorescein-Phalloidin, 300U
Supplier:  Biotium 00030
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. Green fluorescent fluorescein (FITC) phalloidin has excitation/emission at 496/516 nm.
Catalog No. 50-196-4059