Learn More
Enzo Life Sciences Fluorescein Recombinant monoclonal antibody (4-4-20 enhanced) (Human IgG3 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5070200
Encompass_Preferred
Clone: 4-4-20 (enhanced). Isotype: IgG3κ. Immunogen: Fluorescein I-bovine serum albumin (FII-BSA). Applications: Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3033