Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Sodium salt - CAPS Solution, For fluorescence, 95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111004839 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4839 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4839 Supplier MilliporeSigma Supelco Supplier No. 6788410MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

recommended storage temperature 15 C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Percent Purity ≥95.0% (HPLC)
Grade For fluorescence
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.