Learn More
Electron Microscopy Sciences NIGHTSEA™ Leica EZ4 Fluorescence Adapter Mounting Adapter only
SDP
Supplier:  Electron Microscopy Sciences SFAZAD
Encompass_Preferred
Enables use of the NIGHTSEA Stereo Microscope Fluorescence System with the Leica EZ4 series of stereo microscopes, with or without integrated camera. Adapter only.
Catalog No. 50-192-7960