Learn More
Electron Microscopy Sciences NIGHTSEA™ Leica EZ4 Fluorescence Adapter Standard Lamp Base, Violet Barrier Filter

Supplier: Electron Microscopy Sciences SFAZVI
The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts the Leica EZ4 stereo microscope for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between six different excitation/emission combinations and white light to work with a variety of fluorescent proteins and other fluorophores. Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. Includes lamp base with power, lighthead, ring adapter, barrier filter and filter sheild.
Color: Violet
Excitation: 400-415nm
Emission filter: 460 nm longpass
Fluorophores: CFP
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.