Learn More
Electron Microscopy Sciences NIGHTSEA™ Keyence VHX Fluorescence Adapter Best Value UV + VI + RB Full System
SDP
Catalog No. 501930549
Encompass
The NIGHTSEA Model SFA Fluorescence Adapter system adds a versatile fluorescence imaging capability to the Keyence VHX series of digital microscopes. The system is being used successfully by a growing number of Keyence owners for a variety of applications.
Color: UV + VI + RB
Catalog No. 50-193-0549 Supplier Electron Microscopy Sciences Supplier No. SFAKZ2IND
Add to Cart
Add to Cart
Request a Quote
The NIGHTSEA Model SFA Fluorescence Adapter system adds a versatile fluorescence imaging capability to the Keyence VHX series of digital microscopes. The system is being used successfully by a growing number of Keyence owners for a variety of applications.
Color: UV + VI + RB