Learn More
Electron Microscopy Sciences NIGHTSEA™ Fluorescence Adapter Barrier Filter, Ultra Violet

Supplier: Electron Microscopy Sciences SFABFUV
Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing individual light head, barrier filter, and viewing shield combinations. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. The barrier filter clicks on to the ring adapter magnetically, so it is easy to remove it to switch back to white light viewing.
Color: Ultra Violet
Excitation: 360-380nm
Emission filter: 415 nm longpass
Fluorophores: DAPI
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.