Learn More
Sigma Aldrich Fine Chemicals Biosciences Hellma(R) fluorescence cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 10x4 mm, chamber volume 1000 muL | 1EA
Catalog No. NC2183972
Encompass
Hellma(R) fluorescence cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 10x4 mm, chamber volume 1000 muL | 1EA
Catalog No. NC2183972 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8050251EA
Hellma(R) fluorescence cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 10x4 mm, chamber volume 1000 muL | 1EA