Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Fluorescence Detector Performance Standard, 10 mL, Liquid, Fluorescence Detection

Catalog No. 502039953 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This fluorescence detector performance standard is an Alliance HPLC standards kit designed for system assessment. It contains 1 ampule of 10 mL standard solution, which includes 0.10 mg/L Anthracene (± 5%) with a purity of ≥ 99%. The solution is in 70% HPLC Grade Acetonitrile / 30% HPLC Grade Water, volume/volume.

  • Separation mode: Reversed phase
  • Detection method: Fluorescence
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 24 months
  • Shipping condition: Ambient
  • Tolerance: 0.05
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Catalog No. 50-203-9953 Supplier Waters Corp Supplier No. WAT047685
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This fluorescence detector performance standard is an Alliance HPLC standards kit designed for system assessment. It contains 1 ampule of 10 mL standard solution, which includes 0.10 mg/L Anthracene (± 5%) with a purity of ≥ 99%. The solution is in 70% HPLC Grade Acetonitrile / 30% HPLC Grade Water, volume/volume.

  • Separation mode: Reversed phase
  • Detection method: Fluorescence
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 24 months
  • Shipping condition: Ambient
  • Tolerance: 0.05
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.