Learn More
Perkin Elmer US LLC Fluorescence Standard - Rhodamine
Supplier:  Perkin Elmer US LLC L2251366
Encompass_Preferred
The FL 8500 fluorescence spectrometer uses a high-performance continuous wave excitation source with PerkinElmer optics for high sensitivity measurements at scan speeds up to 60,000 nm/min. The FL 8500 is the choice when sensitivity and/or scan speed is most important. The FL 8500 solution can be configured for a range of applications including: material characterization, industrial dye, printing, and tracing manufacutring and R&D, and environmental analysis of LEDs, solar cells and organic electroluminescent materials. The FL 6500 fluorescence spectrometer uses a pulse excitation source with variable power settings desigend for samples susceptible to photo bleaching. The FL 6500 system can be configured for a range of applications including cell biology, molecular biology, immunology, enzymology and protein analysis and thin coatings. Together, these instruments create a comprehensive fluorescence portfolio enabling customers to tackle most application challenges.
Catalog No. 50-211-2783