Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NIGHTSEA™ Stereo Fluorescence Adapter Standard Lamp Base, Royal Blue Barrier Filter
SDP

Catalog No. NC0697697
Encompass_Preferred

The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence w/ no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work w/ a variety of fluorescent proteins & other fluorophores. There are six different excitation/emission combinations available, plus white light.

Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, & bringing fluorescence into the teaching laboratory.

System consists of flexible gooseneck lamp base w/ power supply, light head, ring adapter for microscope, barrier filter, & filter shield.
Color: Royal Blue
Excitation: 440-460nm
Emission filter: 500 nm longpass

Catalog No. NC0697697 Supplier Electron Microscopy Sciences Supplier No. SFARB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence w/ no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work w/ a variety of fluorescent proteins & other fluorophores. There are six different excitation/emission combinations available, plus white light.

Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, & bringing fluorescence into the teaching laboratory.

System consists of flexible gooseneck lamp base w/ power supply, light head, ring adapter for microscope, barrier filter, & filter shield.
Color: Royal Blue
Excitation: 440-460nm
Emission filter: 500 nm longpass

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.