Learn More
Cayman Chemical Fluorescent Brightener 28, CAS: 4193-55-9, 250 mg
SDP
Catalog No. 502943913
Encompass_Preferred
A fluorescent probe; has been used to image chitin in pale grass blue butterfly (Z. maha) wings and β-glucans and glycans in the cell walls of cucumber plant stem and root tissues; displays an emission maximum of 430 nm upon excitation at 360 nm
Catalog No. 50-294-3913 Supplier Cayman Chemical Supplier No. 41995250 MG
A fluorescent probe; has been used to image chitin in pale grass blue butterfly (Z. maha) wings and β-glucans and glycans in the cell walls of cucumber plant stem and root tissues; displays an emission maximum of 430 nm upon excitation at 360 nm