Learn More
Formumax Scientific Inc FLUORESCENT DOPC/CHOL LIPOSOME
Catalog No. NC1488574
Encompass_Preferred
Fluorescent DOPC/CHOL Liposomes labeled with Rhodamine DHPE (100nm, 1.0mL)High quality LUV liposomes labeled with Rhodamine DHPE (Rh-DHPE) are ready to use. The liposomes are prepared by the extrusion method to 100nm with very tight particle size distribution. The product is sterile filtered and filled in sterile vials guaranteed to be stable for 6 months when stored in refrigerator. Rh--DHPE is a phospholipid labeled on the head group with the red-fluorescent rhodamine B fluorophore with excitation/emission maxima ~560/580 nm. For more information on the dye, see Invitrogen L-1392.
Catalog No. NC1488574 Supplier Formumax Scientific Inc Supplier No. F60103FR
Fluorescent DOPC/CHOL Liposomes labeled with Rhodamine DHPE (100nm, 1.0mL)High quality LUV liposomes labeled with Rhodamine DHPE (Rh-DHPE) are ready to use. The liposomes are prepared by the extrusion method to 100nm with very tight particle size distribution. The product is sterile filtered and filled in sterile vials guaranteed to be stable for 6 months when stored in refrigerator. Rh--DHPE is a phospholipid labeled on the head group with the red-fluorescent rhodamine B fluorophore with excitation/emission maxima ~560/580 nm. For more information on the dye, see Invitrogen L-1392.