Learn More
Bachem 5-FAM-Amyloid Beta Protein (1-42) (scrambled) Trifluoroacetate
SDP
Supplier:  Bachem 4099694.0500
Encompass_Preferred
5-FAM-Amyloid Beta Protein (1-42) (scrambled) Trifluoroacetate, Fluorescein-5-carbonyl-Ala-Ile-Ala-Glu-Gly-Asp-Ser-His-Val-Leu-Lys-Glu-Gly-Ala-Tyr-Met-Glu-Ile-Phe-Asp-Val-Gln-Gly-His-Val-Phe-Gly-Gly-Lys-Ile-Phe-Arg-Val-Val-Asp-Leu-Gly-Ser-His-Asn-Val-Ala-OH, , C224H321N55O66S, Mr 4872.41, 0.5mg. 5-FAM-Beta-Amyloid (1-42), Scrambled, Human, Abeta (1-42), Scrambled, FAM labeled, Human. Fluorescent dye-labeled inactive control for amyloid b42.
Catalog No. 50-194-6619