Learn More
Electron Microscopy Sciences Hybriwell™ Fluorescent Friendly 1 well Chamber 90-100 mcL, 0.12 mm
SDP
Catalog No. 5034697
Encompass_Preferred
These new microarray chambers are designed specially for compatibility with Cy5 or Alexa Fluor 647 direct labeled DNA probes for hybridization assays. These chambers as well as the original HybriWells may also be used with all other fluorochrome labeled probes. Each package includes illustrated instructions for use, applicator and 200 adhesive port seals.
Volume: 90-100 µl
I.D.: 22 x 40 mm
Depth: 0.12 mm
Catalog No. 50-346-97 Supplier Electron Microscopy Sciences Supplier No. 7032853
These new microarray chambers are designed specially for compatibility with Cy5 or Alexa Fluor 647 direct labeled DNA probes for hybridization assays. These chambers as well as the original HybriWells may also be used with all other fluorochrome labeled probes. Each package includes illustrated instructions for use, applicator and 200 adhesive port seals.
Volume: 90-100 µl
I.D.: 22 x 40 mm
Depth: 0.12 mm