Learn More
STEMCELL Technologies 2-NBDG, Size: 5 mg
SDP
Catalog No. 502069355
Encompass
2-NBDG is a fluorescent glucose analog and it is used as an indicator for cell viability by monitoring glucose uptake in living cells (Zou et al.). 2-NBDG displays excitation and emission maxima at 465 nm and 540 nm, respectively (Yoshioka et al.). The glucose moiety in 2-NBDG is supplied as a mixture of α and β enantiomers. CANCER RESEARCH· Measures changes in glycolysis and has potential for use in breast cancer cells (Millon et al.).
Catalog No. 50-206-9355 Supplier STEMCELL Technologies Supplier No. 1000547
2-NBDG is a fluorescent glucose analog and it is used as an indicator for cell viability by monitoring glucose uptake in living cells (Zou et al.). 2-NBDG displays excitation and emission maxima at 465 nm and 540 nm, respectively (Yoshioka et al.). The glucose moiety in 2-NBDG is supplied as a mixture of α and β enantiomers. CANCER RESEARCH· Measures changes in glycolysis and has potential for use in breast cancer cells (Millon et al.).