Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroaniline, MilliporeSigma™ Supelco™

Catalog No. 111013994 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3994 25 g
11-101-3993 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3994 Supplier MilliporeSigma Supelco Supplier No. F340125GA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biochem/physiol Actions
The metabolism and excretion of the xenobiotic compound 2-fluoroaniline is important due to human exposure in manufacturing. It is found to be very efficiently metabolized, primarily by 4-hydroxylati

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluoroaniline
Packaging Glass Bottle
CAS 348-54-9
Physical Form Liquid
Linear Formula FC6H4NH2
MDL Number MFCD00007642
Beilstein 1524219
Synonym 1-Amino-2-fluorobenzene
Molecular Weight (g/mol) 111.12
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.