Learn More
Seracare Life Sciences Inc FLUORESCENT MOUNTING MEDIA
Catalog No. 5067564
Encompass_Preferred
Glycerol-based mounting medium for immunofluorescence microscopy. Formulated to retard photobleaching of fluorochromes such as fluorescein and rhodamine. Product can also be used as a diluent for fluorescent-labeled conjugates.
This item is not returnable. View return policy
Catalog No. 50-675-64 Supplier Seracare Life Sciences Inc Supplier No. 55700005
Glycerol-based mounting medium for immunofluorescence microscopy. Formulated to retard photobleaching of fluorochromes such as fluorescein and rhodamine. Product can also be used as a diluent for fluorescent-labeled conjugates.