Learn More
Sigma Aldrich Fluorescent nanodiamond
Catalog No. 9001725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
90-017-25ML 5 mL
1 options
Catalog No. 90-017-25ML Supplier Sigma Aldrich Supplier No. 9001725ML
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Nanodiamonds containing nitrogen-vacancy (NV) centers can be prepared using irradiation with high energy electrons. Nanodiamonds with low numbers of NV centers (ideally, single defect centers) are suitable for quantum signaling, optics, and computing applications because their fluorescence allows for observation of single quantum systems. These particles have approximately 1-4 NV centers per particle as measured using AFM confocal microscopy with subsequent derivation of the correlation function using a Hanbury-Brown-Twiss setup.
Specifications
Quantity 5 mL
Synonym Nanodiamond