Learn More
Sigma Aldrich Fluorescent nanodiamond
Catalog No. 9018005ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
90-180-05ML 5 mL
1 options
Catalog No. 90-180-05ML Supplier Sigma Aldrich Supplier No. 9018005ML
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fluorescent nanodiamond (nitrogen vacancy ∼3 ppm NV centers, 140 nm avg. part. size, carboxylic acid functionalized, PEG 3000 coated, 1 mg/mL in deionized water), a biocompatible carbon-based nanomaterial, has interesting properties like excellent photostability, low toxic and facile surface functionalizability. It can be used for a variety of biological applications.Nanodiamonds (NDs) containing NV color centres exhibit fluorescence in red/near infra-red (NIR) region. The fluorescence in NDs originates from nitrogen vacancies and, unlike quantum dots and organic dyes, NDs do not photobleach. Fluorescent NDs find applications in background-free imaging, flow cytometry, super-resolution imaging etc. Our PEG coated carboxyl functionalized NDs are well suited for attachment of biomolecules using EDC chemistry. The presence PEG spacer on NDs reduces the steric hindrance among the biomolecules.
Specifications
Quantity 5 mL
Synonym Fluorescent nanoparticles; Nanodiamonds