Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fluorescent Particle Array (10 peaks) Kit
SDP

Catalog No. NB397799
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397799 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397799 Supplier Novus Biologicals™ Supplier No. NBP318584

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Used to develop assays for allergy testing, autoimmune diseases, cardiac markers, cytokine detection, endocrine markers, infectious disease markers, isotyping, genotyping, kinase and phosphorylated protein activity, metabolic markers, and tissue typing

Specifications

Content And Storage Store at 4°C. Do not freeze.
For Use With (Application) Flow Cytometry
Format Deionized water
Preservative 0.02% Sodium Azide
Quantity 10 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.