Learn More
Spherotech FLUORESCENT PRTCLS 3.0-3.59UM
Catalog No. NC2222957
Encompass_Preferred
Supra Rainbow Midrange Fluorescent Particles. 10ml, Concentration 106/mL3.0-3.59 µm
This item is not returnable. View return policy
Catalog No. NC2222957 Supplier Spherotech Supplier No. SRCP013510A
Supra Rainbow Midrange Fluorescent Particles. 10ml, Concentration 106/mL3.0-3.59 µm