Learn More
Electron Microscopy Sciences Fluorescent Ruler in inches and centimeters
SDP
Catalog No. 5025972
Encompass_Preferred
Fluorescent Scale in inches and centimeters. This metric/english plastic ruler is printed with a pink phosphorescent ink that glows in the dark. It is most useful for all fluorescent viewing and fluorescent photographic work. The scale will fluoresce visibly upon illumination with ultraviolet light and serves as a measure of nucleic acid mobility. The scale is 0 to 6" with 1/16 " increments on one edge and 0 to 15 cm with 1 mm increments on the other.
Catalog No. 50-259-72 Supplier Electron Microscopy Sciences Supplier No. 6813701
Fluorescent Scale in inches and centimeters. This metric/english plastic ruler is printed with a pink phosphorescent ink that glows in the dark. It is most useful for all fluorescent viewing and fluorescent photographic work. The scale will fluoresce visibly upon illumination with ultraviolet light and serves as a measure of nucleic acid mobility. The scale is 0 to 6" with 1/16 " increments on one edge and 0 to 15 cm with 1 mm increments on the other.