Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Fluorescent Shop Envelopes

Catalog No. 19113411 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Fluorescent Orange Trim on Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-113-411 Fluorescent Orange Trim on Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-113-411 Supplier Brady™ Supplier No. 56944
  • $228.00
    $172.65 / Pack of 25

    Save $55.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color-code work in process, machine procedures and drawings. Brady™ Fluorescent Shop Envelopes are the perfect solution for organizing work flows.

  • Fluorescent yellow trimmed shop envelopes
  • Ideal for color-coding work in process, machine procedures and drawings
  • For indoor and outdoor use

Specifications

Color Fluorescent Orange Trim on Clear
Material Plastic
Mounting Type Metal Grommet
For Use With (Application) Color-coding work in process, machine procedures and drawings
Quantity 25/Pk.
Dimensions (L x W) 9 x 6 in.
Length (English) 9 in.
Width (English) 6 in.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.