Learn More
Electron Microscopy Sciences Gold Seal Fluorescent Antibody Slides, 3 x 1"
SDP
Supplier:  Electron Microscopy Sciences 6373205
Encompass
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage. Frosted one side, one end with white epoxy ink, including circles. 1.2 mm thick with two 10 mm diameter circles.
Size: 3 x 1"
Catalog No. 50-301-68