Learn More
Biophoretics Inc Fluorescent Stain Lava Purple
Catalog No. NC2955639
Encompass
Fluorescent Stain; Lava Purple and Deep PurpleAlso known as ServaPurpleBased on Epicocconone fluorescent stain for proteins and peptides. This fluorescent stain Deep Purple or Serva Purple binds reversibly to lysine, arginine and histidine producing an intense red-fluorescence for detection. Environmentally friendlyEasy to useSensitivity to 50pg/bandLinear response to over 4 orders of magnitudeCompatible with MS, DIGE-labeling and Western blotting
Catalog No. NC2955639 Supplier Biophoretics Inc Supplier No. 43386.01
Fluorescent Stain; Lava Purple and Deep PurpleAlso known as ServaPurpleBased on Epicocconone fluorescent stain for proteins and peptides. This fluorescent stain Deep Purple or Serva Purple binds reversibly to lysine, arginine and histidine producing an intense red-fluorescence for detection. Environmentally friendlyEasy to useSensitivity to 50pg/bandLinear response to over 4 orders of magnitudeCompatible with MS, DIGE-labeling and Western blotting