Learn More
Vector Laboratories Cy5 Sambucus Nigra Lectin (SNL) 1 mg
SDP
Catalog No. NC1487999
Encompass
Sambucus nigra agglutinin (SNA) is a lectin isolated from elderberry bark that binds preferentially to sialic acid attached to terminal galactose in α-2,6 and to a lesser degree, α-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin does not appear to bind sialic acid linked to N-acetylgalactosamine. SNA has been reported to inhibit cell-free protein synthesis. This Cy5-labeled SNA features a ratio of fluorochrome to lectin that provides optimal staining (excitation 649 nm, emission 670 nm, color far red) and is supplied essentially free of unconjugated fluorochrome.
Catalog No. NC1487999 Supplier Vector Laboratories Supplier No. CL13051
Sambucus nigra agglutinin (SNA) is a lectin isolated from elderberry bark that binds preferentially to sialic acid attached to terminal galactose in α-2,6 and to a lesser degree, α-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin does not appear to bind sialic acid linked to N-acetylgalactosamine. SNA has been reported to inhibit cell-free protein synthesis. This Cy5-labeled SNA features a ratio of fluorochrome to lectin that provides optimal staining (excitation 649 nm, emission 670 nm, color far red) and is supplied essentially free of unconjugated fluorochrome.