Learn More
AdipoGen Fluorescin
SDP
Supplier:  AdipoGen CDXD0296G025
Encompass_Preferred
Chemical. CAS 518-44-5. Formula C20 H14 O5. MW 334.32. Synthetic. Fluorescent probe in biology used to detect protein location and activation, identify protein complex formation and conformational changes and monitor biological processes in vitro or in vivo.
Catalog No. 50-256-1140