Learn More
AdipoGen Fluoresdiacetate 5maleimide
SDP
Catalog No. 502560937
Encompass_Preferred
Chemical. CAS 150322-01-3. Formula C28H17NO9. MW 511.4. Synthetic. Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.
Catalog No. 50-256-0937 Supplier AdipoGen Supplier No. CDXF0018M100
Chemical. CAS 150322-01-3. Formula C28H17NO9. MW 511.4. Synthetic. Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.